Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEMIN8 Antibody (OTI1E5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245832
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
GEMIN8 Monoclonal specifically detects GEMIN8 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GEMIN8 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
FAM51A1, FLJ43072, gem (nuclear organelle) associated protein 8, Gemin-8, member A1, Protein FAM51A1 | |
Mouse | |
28.5 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
OTI1E5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
GEMIN8 | |
Full length human recombinant protein of human GEMIN8 (NP_060326) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
54960 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction