Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GEMIN8 Monoclonal Antibody (OTI4F8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526768
Description
GEMIN8 Monoclonal Antibody for Western Blot, IHC (P)
The SMN complex plays a catalyst role in the assembly of small nuclear ribonucleoproteins (snRNPs), the building blocks of the spliceosome. Thereby, plays an important role in the splicing of cellular pre-mRNAs. Most spliceosomal snRNPs contain a common set of Sm proteins SNRPB, SNRPD1, SNRPD2, SNRPD3, SNRPE, SNRPF and SNRPG that assemble in a heptameric protein ring on the Sm site of the small nuclear RNA to form the core snRNP. In the cytosol, the Sm proteins SNRPD1, SNRPD2, SNRPE, SNRPF and SNRPG are trapped in an inactive 6S pICln-Sm complex by the chaperone CLNS1A that controls the assembly of the core snRNP. Dissociation by the SMN complex of CLNS1A from the trapped Sm proteins and their transfer to an SMN-Sm complex triggers the assembly of core snRNPs and their transport to the nucleus.
Specifications
GEMIN8 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NWZ8 | |
GEMIN8 | |
Full length human recombinant protein of GEMIN8 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4F8 | |
Unconjugated | |
GEMIN8 | |
B130034M20Rik; B930082C09Rik; BC023488; Fam51a1; family with sequence similarity 51, member A1; family with sequence similarity 51, member A1 homolog; gem (nuclear organelle) associated protein 8; gem nuclear organelle associated protein 8; gem-associated protein 8; GEMIN8; Gemin-8; Protein FAM51A1 | |
Mouse | |
Affinity chromatography | |
RUO | |
54960 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction