Learn More
Stoelting™ Gemini Active and Passive Avoidance System
Shop All Stoelting Co Products
Click to view available options
Animal:
Mouse
Mouse, Rat
Rat
Model:
Shock
Shock and Additional stations
Shock and Air stimulation
Shock, Air stim., Add'l Stations
Description
- Unique design allows users to run up to 8 independently operated stations directly from a laptop using a standard USB.
- By allowing users to define a complete study in advance, Gemini eliminates run time data entry that could affect the accuracy of results.
- Scrambled shock provides more accurate responses
- Solid state feedback-controlled, constant current shocker uses 8 unique outputs to scramble the shock across the grid floor.
- Scrambling the shock prevents the animal from determining a pattern and leads to more accurate responses.
- Kinder, gentler, unconditioned stimulus - offers an alternative air puff unconditioned stimulus
- Optional Components
- Air puff stimulation unit (AIRSTIM)
- Additional test stations
- Mouse passive avoidance start box
- Grid floors (for rats or mice)
Specifications
Specifications
For Use With (Application) | Neuroscience |
Animal | Mouse |
Model | Shock and Air stimulation |