Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Geminin Antibody (2H7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104293H
Description
Geminin Monoclonal specifically detects Geminin in Human samples. It is validated for Western Blot, ELISA.Specifications
Geminin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
GMNN | |
Recombinant human Geminin (1-209 aa) purified from E. coli | |
0.1 mL | |
Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
51053 | |
Human | |
Purified |
Western Blot, ELISA | |
2H7 | |
Western Blot, ELISA | |
Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human Geminin mAb, clone 2H7, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Geminin protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction