Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Geminin Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10040827APC
Description
Geminin Polyclonal antibody specifically detects Geminin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Geminin | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
Maps to a region between residue 160 and the C-terminus (residue 209) of human Geminin DNA replication Inhibitor using the numbering given in entry NP_056979.1 (GeneID 51053). | |
0.1 mL | |
Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
51053 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction