Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Geminin Antibody (GMNN/7037R), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314129JF549
This item is not returnable.
View return policy
Description
Geminin Monoclonal antibody specifically detects Geminin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Geminin | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
GMNN/7037R | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
Recombinant fragment (around aa71-202) of human Geminin protein (exact sequence is proprietary) (Uniprot: O75496) | |
0.1 mL | |
Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
51053 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction