Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Geminin Mouse anti-Human, Clone: GMNN/3665, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Geminin |
---|---|
Clone | GMNN/3665 |
Concentration | 0.2 mg/ml |
Dilution | Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
Description
Geminin Monoclonal antibody specifically detects Geminin in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Geminin | |
0.2 mg/ml | |
Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA Replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
Recombinant fragment of human Geminin protein (exact sequence is proprietary) (Uniprot: O75496) | |
Primary | |
Store at 4°C. |
GMNN/3665 | |
Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
O75496 | |
51053 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title