Learn More
Geminin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | Geminin |
|---|---|
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-8833
|
Bethyl Laboratories, Inc
A300935AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The DNA replication (origin) licensing system is necessary for duplicating the genome in each cell cycle. Geminin is a protein that prevents DNA replication by blocking the MCM complex incorporation into prereplication complex (pre-RC). It is absent during G1 phase and accumulates during S, G2, and M phases of mitosis. It is then degraded during the metaphase-anaphase trasition allowing replication to occur during subsequent cycles. Geminin has also been implicated in maintaining integrity of the male germline genome.Specifications
| Geminin | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Gem, geminin | |
| GMNN | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75496 | |
| 51053 | |
| Between 160 and C-term | |
| Primary | |
| 4° C | |
| GMNN |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.