Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemtuzumab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541705
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541705 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541705 Supplier Invitrogen™ Supplier No. MA541705
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Gemtuzumab ozogamicin is a recombinant humanized IgG4 kappa antibody which is conjugated with calicheamicin derivative, a cytotoxic antitumor antibiotic isolated from fermentation of Micromonospora echinospora ssp. calichensis. Gemtuzumab ozogamicin has approximately 50% of the antibody loaded with 4-6 moles calicheamicin per mole of antibodyLabel. The antibody is specifically directed against the CD33 antigen present on leukemic myeloblasts in most patients with acute myeloid leukemia (AML). By binding to the CD33 antigen on tumors, the cytotoxic agent blocks the growth of cancerous cells and causes cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemtuzumab Humanized
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 3.38 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias CDP-771; CMA-676 (ADC); gemtuzumab ozogamicin; hP67.6; hP67.6-calicheamicin (ADC); WAY-CMA-676
Host Species Human
Immunogen Human CD33.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG4 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.