Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GEN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554137 Supplier Invitrogen™ Supplier No. PA554137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SILSVKESCI ANSGSDCTSH LSKDLPGIPL QNESRDSKIL KGDQLLQEDY KVNTSVPYSV SNTVVKTCNV RPPNTALDHS RKVDMQTTRK ILMKKSVCLD RHSSDEQSAP VFGKAKYTTQ RMKHSSQKHN SSHFKESGHN KLSSPKI Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 47%.

Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEN1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GEN1
Gene Accession No. Q17RS7
Gene Alias 5830483C08Rik; Flap endonuclease GEN homolog 1; Gen; Gen endonuclease homolog 1; Gen homolog 1, endonuclease; Gen homolog 1, endonuclease (Drosophila); GEN1; GEN1 Holliday junction 5' flap endonuclease; GEN1, Holliday junction 5' flap endonuclease; Holliday junction resolvase; RGD1559792
Gene Symbols GEN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GEN1. Recombinant protein control fragment (Product #RP-92437).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.