Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEN1 Rabbit anti-Human, Rat, Clone: S09-1B8, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $586.50
Specifications
Antigen | GEN1 |
---|---|
Clone | S09-1B8 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
GEN1 Monoclonal antibody specifically detects GEN1 in Human, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
GEN1 | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
DKFZp781F0986, EC 3.1, flap endonuclease GEN homolog 1, FLJ40869, Gen, Gen homolog 1, endonuclease (Drosophila) | |
A synthetic peptide of human GEN1 (Uniprot#: Q17RS7) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S09-1B8 | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Direct Reversal of DNA Damage | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
348654 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title