Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ Gibson Assembly EX Master Mix

Catalog No. p-7181131
Encompass_Preferred
Click to view available options
Quantity:
10 Reactions
200 Reactions
50 Reactions

GeneArt Gibson Assembly EX Master Mix enables DNA assembly via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a single-tube, two-step optimized reaction.

GeneArt Gibson Assembly EX Master Mix enables DNA assembly via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a single-tube, two-step optimized reaction. DNA fragments of different lengths are uniformly assembled using complementary overlaps between fragments. The inherent flexibility and success of this approach is suitable for small and large DNA constructs, includes both single and multiple inserts, and has been used to build entire genomes. The resulting products can be used for a variety of downstream applications, including transformation, PCR, and rolling circle amplification (RCA). The GeneArt Gibson Assembly EX Master Mix kit includes master mix, positive control, and water, and accommodates the use of your own competent cells.

Features of the GeneArt Gibson Assembly EX Master Mix include:

  • Simple—seamlessly assemble and clone up to 15 DNA fragments in a single reaction
  • Efficient—robust efficiency provides successful clones for both simple and challenging constructs
  • Flexible—design guidelines allow assembly into any vector of your choice
  • Convenient—available as master mix kits and cloning kits complete with chemical or electrocompetent cells
  • Trusted—over 4000 citations in the scientific literature highlighting great success

Seamless assembly of up to 15 fragments

GeneArt Gibson Assembly EX cloning is a robust, single-tube, two-step process whereby up to 15 inserts and vector are combined in a proprietary enzymatic mix in order to assemble DNA fragments with shared terminal end homology without leaving any extra sequences or scars behind (seamless). After this dually optimized reaction is complete, a portion of the assembly reaction is then transformed into chemically competent or electrocompetent cells, yielding clones ready for analysis the next day. The required 20- to 40-base pair end homology is designed into the de novo fragment for synthesis or can be easily engineered by PCR amplification with custom DNA oligos. This special enzyme mix creates a seamless and covalently bound DNA construct providing high efficiency.

Robust method for maximum efficiency

Due to the covalently bound final product, the GeneArt Gibson Assembly EX method allows the utilization of chemically competent cells or electrocompetent cells for the highest transformation efficiency, especially important for challenging constructs. There is no need for restriction enzymes, ligation, or recombination sites, and the method provides a perfectly seamless construct without unwanted extra bases.

Provides great versatility

The GeneArt Gibson Assembly EX method provides versatility, can streamline many techniques through the rapid combination, addition, deletion, or exchange of DNA segments, and eliminates the need to subclone, saving time and effort in the cloning workflow.

TRUSTED_SUSTAINABILITY

Specifications

Cloning Method Seamless cloning
Content And Storage Includes master mix, positive control, and water and accommodates the use of your own competent cells.

Control Type Positive Control
For Use With (Application) Seamless Cloning & Genetic Assembly
No. of Reactions 10 Reactions
Number of Fragments Up to 15 fragments
Product Line GENEART™
Product Type Master Mix
Quantity 10 Reactions
Shipping Condition Dry Ice
Workflow Step DNA Assembly
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.