Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ Site-Directed Mutagenesis PLUS System

Catalog No. a14604
Encompass_Preferred
Click to view available options
Quantity:
10 Reactions

Brings our mutagenesis technology to the next level with the ability to perform single- or multi-site mutations in larger plasmids with greater efficiency and in less time than the competition

Features:
  • Powerful—make substitutions, deletions, or insertions of up to 3 nucleotides in 1, 2, or 3 separate sites in DNA plasmids of up to 14kb
  • Flexible—perform multi-site mutagenesis with degenerated nucleotides and single-site mutations up to 25 nucleotides long or 12 nucleotides long with degenerated nucleotides
  • Efficient—obtain your desired mutants the first time; over 90% correct mutants in a 10kb plasmid
  • Fast—obtain your mutated plasmid DNA in typically less than 3 hours with the simple, minimal-handling protocol
  • Versatile—use plasmids of many sizes, and DNA isolated from any source, with no need for specialized vectors, host strains, or restriction sites

Optimized Mutagenesis Protocol
The GeneArt™ Site-Directed Mutagenesis System was further optimized for efficiency and multi-site capability, resulting in the “PLUS” kit. Like the first generation GeneArt™ Site-Directed Mutagenesis System, DNA methylation and amplification steps are combined into a single reaction, with no need for an in vitro DpnI digestion step. After methylation and amplification, a 15 minute in vitro recombination reaction of the amplified PCR products increases the colony output by 3 to 10 fold, resulting in a higher mutagenesis efficiency. A final transformation of the mutated DNA into DH5α™ E.coli cells digests any methylated parental DNA, leaving behind only the intact unmethylated mutagenesis reaction product. No purification steps or additional digestions are needed. Individual colonies can be selected the following day to verify mutations.

Simple Creation of Desired Mutants
Creating mutants with the GeneArt™ Site-Directed Mutagenesis PLUS System relies on the inherent properties of DNA methylase, high fidelity DNA polymerase, recombination enzymes, and the native McrBC endonuclease of E. coli. Simply incorporate your desired mutation or mutations into primers using our GeneArt™ Design Tool for Seamless Assembly and Mutagenesis (see “in silico Design Support”, below). Combine the vector and mutagenesis primers, and after PCR, recombination, and transformation you will have vectors with only the desired mutations with over 90% efficiency.

in silico Design Support
A key step in GeneArt™ site-directed mutagenesis is the correct design of primers with the appropriate homology and spacing to help ensure successful mutagenesis of your plasmid. To simplify and speed the design process, we provide a free online tool, the GeneArt™ Design Tool for Seamless or High-Order Assembly and Mutagenesis, to help you design your experiment in silico. The tool allows you to review your plasmid sequence, designs DNA oligos for introduction of the desired mutations, provides an updated amino acid sequence for your mutated gene, displays a graphical representation of the new construct, and enables the download of an annotated sequence in GenBank format that is compatible with Vector NTI™ software.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain DH5α
Cell Type Chemically Competent E. coli
Content And Storage • GeneArt 2X Enzyme Mix (12 μL)
• One Shot MAX Efficiency DH5α-T1(1 box)
• DNA Methylase (12 μL)
• 200X SAM (10 μL)
• 10X PCR Enhancer (100 μL)
• PCR Water (1.8 μL)
• pMSDM-White Vector (5 μL)
• Control Primer Mix 1, 2, & 3 (20 μL each)
• 0.5 M EDTA (500 μL)

For convenience, all contents may be stored at −80°C.
Efficiency 90 %
For Use With (Application) Seamless Cloning & Genetic Assembly
Mutagenesis Capability Mutagenesis at Multiple Sites
Mutagenized Region Length (up to) 3 nucleotides (multi-site), 25 nucleotides (single-site)
Mutation Type Deletions, Insertions, Substitutions
Plasmid Plasmids up to 14 kb
Product Type Site-Directed Mutagenesis PLUS System
Quantity 10 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.