Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ Seamless Cloning and Assembly Enzyme Mix

Catalog No. a14606
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14606 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14606 Supplier Invitrogen™ Supplier No. A14606
Only null left
Add to Cart
Edge
Add to Cart

Enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute room temperature reaction

GeneArt™ Seamless Cloning and Assembly Enzyme Mix enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute room temperature reaction. GeneArt™ Seamless enzyme mix is the economical choice for creating constructs up to 13 kb with the option for high-throughput assembly.
  • Efficiency: pre-cloning option for large fragments for increased cloning efficiency
  • Speed and Ease: clone DNA fragments, with sequence of your choice, in a single vector (up to 13 kb); no restriction digestion, ligation, or recombination sites required
  • Free Tools: design of your final construct and DNA oligos in silico using our free web-based tool that takes you step-by-step through your project
  • Vector Flexibility: use our linear vector or a vector of your choice

For the cloning of pre-existing DNA elements too long to be amplified by PCR and for final molecules up to 40 Kb, please consider the GeneArt™ Seamless PLUS Cloning and Assembly Kit (1–4 DNA fragments only); and for final molecules larger than 110 Kb, please consider the GeneArt™ High-Order Genetic Assembly System.

Simple and Fast Clone Creation
GeneArt™ Seamless Cloning is a simple, two step process, consisting of in vitro assembly followed by transformation. The enzyme mix uses a proprietary enzyme/buffer mix to assemble DNA fragments without extra sequences or scars (“seamless”). Terminal end homology, if needed, is easily incorporated by PCR amplification with custom DNA oligos engineered using our free web tool.

Cloning Efficiency, Flexibility, and Precision
Cloning success is independent of the insert sequence and vector type, allowing you to design and add nearly any desired sequence, or combination of sequences, to any plasmid as long as it can be linearized by either restriction enzyme digestion or PCR. The circularized clones obtained from the reaction contain only the sequence of your original vector, inserts, and designated homologies, with no extraneous nucleotide insertions.

in silico Cloning Design Support
A key step in GeneArt™ Seamless Cloning is the correct design of fragments and oligos with the appropriate homology and spacing to help ensure successful assembly of your clone. To simplify and speed the design process, we provide a free online tool, the GeneArt™ Design Tool for Seamless or High-Order Assembly and Mutagenesis, to help you design your experiment in silico. The tool guides the user through construct design, including fragment import, order, and orientation; checks for areas of homology and potential design issues; creates primers for pre-cloning or incorporating end homology between fragments, if needed; and generates final construct map files and sequence in Genbank format for download or import into Vector NTI™ software.

Applications
The GeneArt™ Seamless Cloning and Assembly Enzyme Mix is designed to empower cloning and DNA assembly in a wide range of molecular biology and synthetic biology applications, among others. The product allows for the creation of modular expression vectors, with interchangeable parts, and can be used to perform a variety of tasks that would otherwise involve multiple steps. Use the mix to construct fusion proteins; delete, replace, or add DNA elements such as restriction sites in an existing vector; and carry out many other techniques that require manipulation of genetic sequences.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY

Specifications

Cloning Method Seamless Cloning
Content And Storage 1 vial GeneArt Enzyme Mix (2X)
pUC19L Linearized Vector
Control Insert
Format Kit
For Use With (Application) Cloning
Number of Fragments Up to 4 Fragments
Quantity 20 Reactions
Size 13 kb total (vector plus all inserts)
Vector pUC19L Linearized
Workflow Step DNA Assembly

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.