Learn More
Thermo Scientific™ GeneBLAzer™ FXR-UAS-bla HEK 293T Cells
Description
GeneBLAzer™UASbla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-FXR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. FXR-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of chenodeoxycholic acid (CDCA). In addition, FXR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | Shipping: Dry IceStorage: -80°C/Liquid NitrogenContains: FXR-UAS-bla HEK 293 T cells (K1239) |
| Detection Method | Fluorescence |
| Druggable Target | Nuclear Receptors |
| Format | 384-well plate, Tube |
| Product Line | GeneBLAzer |
| Product Type | Cell Line |
| Quantity | 2 x 106 cells |
| Reporter Gene | BLA (Beta-Lactamase) |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.