Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ RARα-UAS-bla HEK 293T Cells

Catalog No. K1685
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1685 2 x 106 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1685 Supplier Thermo Scientific™ Supplier No. K1685
Only null left
Add to Cart
Add to Cart
Request a Quote

RAR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor alpha fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK293T cell line.

RAR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor alpha fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK293T cell line. GeneBLAzer™UAS-bla HEK 293T cells (Cat. No. K1104) stably express a beta-lactamase reporter gene under the transcriptional control of a 7X Upstream Activator Sequence (UAS). Transcription from the 7X UAS is activated by the binding of the GAL4 transcription factor DNA-binding-domain (DBD). The GAL4-DBD is expressed as a fusion protein with the ligand binding domain (LBD) of RAR alpha. When an agonist binds to the LBD of the GAL4(DBD)-RAR alpha(LBD) fusion protein it translocates to the nucleus where it binds to the 7X UAS inducing transcription of betalactamase. RAR alpha-UAS-bla HEK 293T cells have been tested for assay performance using variable assay conditions, including DMSO concentration, cell number, stimulation time, substrate loading time and have been validated for Z' and EC50 concentrations of all-trans retinoic acid. Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists Tetrac, T4
Antagonists 1-850
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage TR alpha-UAS-bla HEK 293T cells (K1260)

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Detection Method Fluorescence
Druggable Target Nuclear Receptors
No. of Cells 2 x 106 Cells
Primary Agonist T3 Hormone
Product Line GeneBLAzer
Product Type HEK 293T Cells
Quantity 2 x 106 cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry RARA, RAR alpha, NR1B1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.