Learn More
Thermo Scientific™ GeneJET Gel Extraction Kit PROMO
Description
Thermo Scientific GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.
The kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column. The kit can be used to purify DNA fragments from 25 bp to 20 kb in size with recovery rates up to 95%. Each GeneJET purification column has a binding capacity of up to 25 μg of DNA and can process up to 1 g of agarose gel. The entire procedure takes just 15 minutes, and the isolated DNA is ready to use in all common downstream applications, including ligation, restriction digestion, PCR, sequencing, and labeling.
Highlights
- Highly efficient: up to 95% recoveries in the range of 100 bp to 10 kb
- Fast: procedure takes only 15 minutes
- Convenient: spin columns are capped and assembled with collection tubes
Applications
Fast and efficient extraction of high purity DNA fragments ideal for use in all conventional molecular biology procedures including:
- FastDigest or conventional restriction digestion
- Automated fluorescent or radioactive sequencing
- PCR
- In situ hybridization
- Labeling
- Cloning
Specifications
Specifications
| Column Type | Spin Column |
| Content And Storage | 30 mL Binding Buffer, 9 mL Wash Buffer (concentrated), 15 mL Elution Buffer, 50 GeneJET Purification Columns with Collection Tubes |
| Isolation Technology | Silica Spin Column |
| Sample Type | Agarose gels |
| Elution Volume | 50 μL |
| Final Product Type | DNA |
| For Use With (Application) | PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Quantity | 250 Preps |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.