Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneJET™ Gel Extraction Kit PROMO

Catalog No. ferk0691pm
Click to view available options
:
For 50 preps

Catalog No. FERK0691PM

Supplier: Thermo Scientific™ K0691

Thermo Scientific™ GeneJET™ Gel Extraction Kit allows one to perform rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.

The kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column. The kit can be used to purify DNA fragments from 25 bp to 20kb in size with recovery rates up to 95%. Each GeneJET purification column has a binding capacity of up to 25μg of DNA and can process up to 1g of agarose gel. The entire procedure takes just 15 min. and the isolated DNA is ready to use in all common downstream applications including ligation, restriction digestion, PCR, sequencing and labeling.
  • Highly efficient: up to 95% recoveries in the range of 100 bp—10kb

  • Fast: procedure takes only 15 minutes

  • Convenient: spin columns are capped and assembled with collection tubes

Recommended for:

Fast and efficient extraction of high purity DNA fragments ideal for use in all conventional molecular biology procedures including:—FastDigest or conventional restriction digestion;—automated fluorescent or radioactive sequencing

Specifications

For Use With (Application) Fast and efficient extraction of high purity DNA fragments ideal for use in all conventional molecular biology procedures including:—FastDigest or conventional restriction digestion;—automated fluorescent or radioactive sequencing
No. of Reactions 50 Preps
Product Type GeneJET Gel Extraction Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.