Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneJET PCR Purification Kit PROMO

GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Supplier:  Thermo Scientific™ K0702

 View more versions of this product

Catalog No. FERK0702PM


Description

Description

Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions. It effectively removes primers, dNTPs, unincorporated labeled nucleotides, enzymes, and salts from PCR and other reaction mixtures.

The kit can be used for purification of DNA fragments from 25 bp to 20 kb with recovery rates up to 100%. Each GeneJET purification column has a total binding capacity of up to 25 μg of DNA, and the entire procedure takes 5 minutes. The purified DNA can be used in common downstream applications, such as sequencing, restriction digestion, labeling, ligation, cloning, in vitro transcription, blotting or in situ hybridization.

Highlights

  • Highly efficient: 90 to 100% recoveries in the range of 100 bp to 10 kb
  • Fast: procedure takes only 5 minutes
  • Convenient: spin columns are capped and assembled with collection tubes

Applications

  • Fast and efficient purification of DNA fragments ideal for use in all conventional molecular biology procedures including:
  • FastDigest or conventional restriction digestion
  • Automated fluorescent or radioactive sequencing
  • PCR
  • In vitro transcription
  • In situ hybridization
  • Labeling
  • Cloning
Specifications

Specifications

60 mL Binding Buffer, 45 mL Wash Buffer (concentrated), 30 mL Elution Buffer, 250 GeneJET Purification Columns with Collection Tubes
50 μL
5 min.
5 min.
≤400 μL
Nucleic Acid Labeling, Southern Blotting, DNA Sequencing, Nucleic Acid Hybridization
Silica Spin Column
Spin Column
Not High-throughput Compatible (Manual)
250 Preps
Room Temperature
25 μg (Binding capacity)
DNA
PCR Reactions, Enzymatic Reactions (DNase digestions, RNA labeling and clean-up preps, IVT reactions)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.