Learn More
Thermo Scientific™ GeneJET RNA Purification Kit PROMO
Description
Thermo Scientific GeneJET RNA Purification Kit is a simple and efficient system for purification of total RNA from mammalian cultured cells, tissue, human blood cells, bacteria, and yeast.
The kit utilizes a silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious cesium chloride gradients, alcohol precipitation, or toxic phenol-chloroform extractions.
RNA molecules longer than 200 nucleotides can be isolated with the GeneJET RNA Purification Kit in 15 minutes after the lysis step. The high-quality purified RNA can be used in a wide range of downstream applications, including RT-PCR, RT-qPCR, Northern blotting, and other RNA-based analyses.
Highlights
- Universal–can be used for both cell and tissue samples from a wide range of sources: mammalian blood, mammalian cell cultures, mammalian tissues, insect, yeast, and bacteria
- Efficient–high yields of total RNA.
- Fast–15 minutes after lysis step
- Pure–an A260/280 ratio > 1.9
- Convenient–spin columns are capped and assembled with collection tubes
Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
- RT-PCR
- RT-qPCR
- Northern blotting
- Nuclease protection assay
Includes
Proteinase K Lysis Buffer, Wash Buffer 1 (concentrated), Wash Buffer 2 (concentrated), Nuclease-free Water, GeneJET RNA Purification Columns (pre-assembled with Collection Tubes), Collection Tubes (2 mL), Collection Tubes (1.5 mL), Detailed Protocol
Specifications
Specifications
| Content And Storage | 600 μL Proteinase K, Store at room temperature until opened, -20°C after opening, 40 mL Lysis Buffer, 40 mL Wash Buffer 1 (concentrated), 23 mL Wash Buffer 2 (concentrated), 30 mL Nuclease-free Water, 50 GeneJET RNA Spin Columns with Collection Tubes, 50 Collection Tubes (2 mL), 50 Collection Tubes (1.5 mL) |
| Isolation Technology | Silica Spin Column |
| Purification Time | 15 min. (After sample homogenization) |
| Elution Volume | 50 to 100 μL |
| Sample Type | Bacteria, Blood, Cells, Tissue, Yeast, Insects, Enzymatic Reactions |
| Final Product Type | Total RNA |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| For Use With (Application) | RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling |
| Quantity | 50 Preps |
| Scale | Mini |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.