Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeneMapper™ ID-X Software v1.6, client installation

Catalog No. A39976
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 License
10 License
5 Licenses
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39976 1 License
A39977 5 Licenses
A39978 10 License
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. A39976 Supplier LIFE TECHNOLOGIES Supplier No. A39976
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Product Line GeneMapper
Content And Storage Store at room temperature.
Product Type GeneMapper ID-X Software V1.6 Client Installation
Quantity 1 License
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.