Learn More
Thermo Scientific™ General Purpose Centrifuge Rotor Packages

Description
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Material | See individual centrifuge, rotor and accessory data |
Type | Rotor Equipment Package |
Capacity | Max in TX-1000: 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
Net Weight | See individual components of package |
For Use With (Equipment) | Purchase of Thermo Scientific General Purpose Centrifuge required for operation |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 |
Includes | TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters |
Warranty | See Manual |
Biocontainment | Yes |
Model | TX-1000 Blood Tube Package |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.