Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ General-Purpose Chromatography columns

Catalog No. 111010453 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
12.7 cm
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Volume (Metric)
11-101-0453 12.7 cm 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-0453 Supplier MilliporeSigma™ Supelco™ Supplier No. C2103200EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in general chromatographic work for extraction of components, with special applications in isoenzyme and protein studies, hemoglobin fractionation and desalting.

Precision molded from premium-grade, chemically inert plastic for organic solvents. Includes reservoir and end caps. Polyethylene filter matrix support in medium porosity.
TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 12.7 cm
Volume (Metric) 10 mL
Column Type Chromatography
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.