Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneRuler 100 bp DNA Ladder PROMO

Catalog No. FERSM0242PM
Click to view available options
Quantity:
5 x 50 μg

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.

The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 100 bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase, protocol. The ladder is supplied with 6X DNA Loading Dye.

  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Product Type DNA Ladder
Ready to Load No
Size Range 100 to 1000 bp
Gel Compatibility Agarose Gels
Product Line GeneRuler
Quantity 5 x 50 μg

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.