Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories

Catalog No. 14380452
Click to view available options
Type:
Dust Cover
Replacement Compartment Lid
Self-Stick Printer Paper
Standard Printer Paper
Thermal Snap-on Printer
Tungsten Halogen Lamp

Catalog No. 14-380-452

Supplier: Thermo Scientific™ 840278400

Only null left
Add to Cart
Add to Cart

Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.

Protects instrument while not in use. Protects against dust and any liquid exposure
TRUSTED_SUSTAINABILITY

Specifications

Type Dust Cover
For Use With (Application) Keep dust and spills off your spectrophotometer when it is not in use
For Use With (Equipment) GENESYS 30 Visible Spectrophotometer
Includes 1 dust cover
Storage Requirements Ambient
Warranty 2 Years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.