Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GENESYS™ 30 Dust Cover
SCI_ED

Catalog No. s28523
Click to view available options
For Use With (Equipment):
GENESYS 30 Visible Spectrophotometer
Includes:
1 dust cover

Our dust cover keeps your GENESYS 30 safe while you're not using it. When you need to use it again just remove the cover and you'll be on your way.

Protects instrument while not in use. Protects against dust and any liquid exposure
TRUSTED_SUSTAINABILITY

Specifications

Description Transparent plastic dust cover protects instrument against dust and any liquid exposure while not in use.
For Use With (Application) Keep dust and spills off your spectrophotometer when it is not in use
For Use With (Equipment) GENESYS 30 Visible Spectrophotometer
Includes 1 dust cover
Storage Requirements Ambient
Warranty 2 Years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.