Learn More
Thermo Scientific™ GENESYS™ 30 Standard Printer Paper
SCI_ED
Catalog No. S28520
Change view
Click to view available options
Includes:
10 rolls of paper
Catalog No. Includes
S28520 10 rolls of paper
1 options
This item is not returnable. View return policy
Catalog No. S28520 Supplier Thermo Scientific™ Supplier No. 840278300/EMD
This item is not returnable. View return policy
Printer paper made just for the GENESYS 30 makes reporting your data fast and easy.
Package of 10 rolls
80mm width x 50mm roll diameter
typical legibility = 5 years
TRUSTED_SUSTAINABILITY
Specifications
Description Standard Printer Paper
For Use With (Equipment) Use with the printer for GENESYS spectrophotometers
Includes 10 rolls of paper