Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genethonin 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982812
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2812 Supplier Bioss Supplier No. BS13335R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May have the capability to bind to carbohydrates.

Specifications

Antigen Genethonin 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene STBD1
Gene Accession No. O95210
Gene Alias D530019K15Rik; D5Ertd593e; genethonin 1; Genethonin-1; GENEX3414; GENX-3414; Glycophagy cargo receptor stbd1; starch binding domain 1; Starch-binding domain-containing protein 1; Stbd1
Gene Symbols STBD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Genethonin 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8987
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.