Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genethonin 1 (STBD1) Monoclonal Antibody (OTI6E9), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene CF812279

Encompass
Encompass_Preferred

Catalog No. 50-254-3726


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Genethonin 1 (STBD1) Monoclonal antibody specifically detects Genethonin 1 (STBD1) in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Genethonin 1 (STBD1)
Monoclonal
See Label
PBS with 8% trehalose and no preservative; pH 7.3
Q14134, Q8R2Q0
Mouse
Protein A/G
RUO
23650, 300656, 72169
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI6E9
Unconjugated
Trim29
D530019K15Rik; D5Ertd593e; genethonin 1; genethonin-1; GENEX3414; GENX-3414; Glycophagy cargo receptor stbd1; RGD1311800; starch binding domain 1; starch-binding domain-containing protein 1; Stbd1
Human recombinant protein fragment corresponding to amino acids 1-352 of human TRIM29 (NP_036233) produced in E.coli.
100 μg
Primary
Human, Mouse, Rat
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.