Learn More
Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder
Description
Geneticin™ Selective Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells. Resistance to Geneticin™ Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3') – I and APH (3') – II resistance genes. Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) is used as a selective antibiotic in the concentration range of 100 - 200 μg/mL for bacteria, or 200 – 500 μg/mL for most mammalian cells. This product is supplied as a powder and should be made into a stock solution of 10 – 50 mg/mL in water. We offer a variety of cell culture antibiotics for your convenience.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Dual site cGMP Manufacturing and Quality System
For supply chain continuity, we manufacture Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Content And Storage | Storage conditions: 15 to 30°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
| Cell Type | Eukaryotic Cells, Prokaryotic Cells |
| Concentration | 10 to 50 μg/mL |
| Culture Type | Mammalian Cell Culture, Insect Cell Culture |
| Form | Powder |
| Product Type | Antibiotic |
| Sterility | Sterile |
| For Use With (Application) | Eukaryotic Selection/Stable Cell Line Generation |
| Product Line | Geneticin |
| Quantity | 25 g |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.