Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genistin, 99+%, Thermo Scientific Chemicals

Catalog No. aaj63445mc
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63445MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63445MC Supplier Thermo Scientific Chemicals Supplier No. J63445MC
Only null left
Add to Cart

Glucoside of genistein that inhibits protein tyrosine kinase

Glucoside of genistein that inhibits protein tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-59-9
Molecular Formula C21H20O10
Molecular Weight (g/mol) 432.381
MDL Number MFCD00016883
InChI Key ZCOLJUOHXJRHDI-CMWLGVBASA-N
Synonym genistin, genistine, genisteol 7-monoglucoside, genistoside, genistein 7-glucoside, glucosyl-7-genistein, genistein 7-o-beta-d-glucoside, unii-1pog3scn5t, genistein glucoside, genistein-7-glucoside
PubChem CID 5281377
ChEBI CHEBI:27514
IUPAC Name 5-hydroxy-3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=CC(=CC(=C3C2=O)O)OC4C(C(C(C(O4)CO)O)O)O)O

Specifications

Melting Point 258°C to 259°C
Color Brown to Yellow
Quantity 100 mg
Beilstein 64479
Merck Index 14,4391
Formula Weight 432.38
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Genistin
RTECSNumber DJ3093000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.