Learn More
Amsbio LLC (formerly Genlantis) BL21 Gen-X™ Competent E. coli Expression Kit
Click to view available options
Quantity:
1 Ea.
Description
Includes
Kit includes BL21 Gen-X Chemically Competent E. coli, 10 x 50μL; SOC medium, 3mL; pUC19 positive control plasmid, 20μL (500pg/μL); Gen-X Induction Enhancer, 2 x 10mL (10X).
- Up to 10X greater expression of recombinant proteins
- Reduced background protein levels, as much as several fold
- Detect proteins not visible if expressed in other BL21 strains
- Simple, straightforward protocol
- Compatible with all T7 promoter-based expression vectors
Specifications
Specifications
For Use With (Application) | Provides significantly higher levels of recombinant Protein expression and significantly reduced levels of background Proteins when compared to previous generations of BL21(DE3) |
Includes | 10 x 50μL BL21 Gen-X chemically competent E. coli, 3mL SOC medium, 20μL (500pg/μL) pUC19 positive control plasmid, 2 x 10mL (10X) Gen-X induction enhancer |
Quantity | 1 Ea. |