Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amsbio LLC (formerly Genlantis) Detachin™ Cell Detachment Reagent
Click to view available options
Quantity:
100 mL
6 x 50mL
Description
- Maximum cell viability
- Effective on a wide range of cells
- No mammalian or bacterial byproducts
- Stable at 4°C for two months
- No need to wash detached cells
- Suitable for detaching adherent cells from in vitro growth vesicles
- Provides quick and gentle detachment and dissociation of a wide variety of adherent cells, including primary cells, grown in all known tissue culture plasticware
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | Cell Culture Dissociation Reagent |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction