Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genlantis™ MycoScope™ PCR Detection Kit

Catalog No. MY01050
Encompass
Change view
Click to view available options
No. of Reactions:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MY01050 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MY01050 Supplier Genlantis™ Supplier No. AMS.MY01050

Includes

dNTP/Buffer mixture (1X); Primer mix (5X); Positive control reaction tube; Molecular biology grade water

Designed for sensitive detection of mycoplasma using PCR

  • Allows detection of less than five mycoplasma genomes/μL sample
  • Unique primer sets detect all mycoplasma species commonly encountered in cell culture
  • Fast and easy protocol eliminate the need for labeling probes

Specifications

Content And Storage 2 x 1.25mL dNTP/Buffer mixture 1X, 530μL primer mix 5X, 50μL positive control reaction tube, 1.5mL molecular biology grade water. Store at -20°C
For Use With (Application) Microbial Detection
No. of Reactions 50 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.