Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amsbio LLC (formerly Genlantis) SoluLyse™ Bacterial Protein Extraction Reagent

Catalog No. p-3545097
Encompass
Encompass_Preferred
Click to view available options
Quantity:
125 mL
500 mL
Formulation:
20mM Tris HCl Buffer, pH 7.5
50mM MNaHPO4, pH 7.4

Efficient and gentle extraction of soluble proteins from bacteria

  • Produces high yield of soluble proteins
  • Simple one-step protocol
  • Proprietary formulation of nonionic detergents optimized for bacterial protein extraction
  • Proteins retain more activity
  • Compatible with protease inhibitors, salts, chelating agents and reducing agents
  • Choice of Phosphate or Tris buffer formulations to avoid interference with affinity purification resins and other downstream applications

Specifications

Formulation 20mM Tris HCl Buffer, pH 7.5
Quantity 125 mL
Product Type Bacterial Protein Extraction Reagent
For Use With (Application) Proprietary formulation of nonionic detergents optimized for bacterial Protein extraction, high Protein yield with simple one-step proto col
Content And Storage Room Temperature
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.