Learn More
Amsbio LLC (formerly Genlantis) Subculturing Reagents Kit
Catalog No. pr090100k
Encompass
Encompass_Preferred
Click to view available options
Quantity:
1 Kit
Includes
PR090100K: Hank's Buffered Saline Solution, 100mL; Trypsin/EDTA, 100mL; Trypsin Neutralizing Solution, 100mL
Functionally tested subculturing reagent for primary cells
  • Includes 100mL each of Hanks Buffered saline, Trypsin/EDTA, and Trypsin Neutralizing solutions
  • Individual reagents also available
Specifications
Product Type Subculture Reagent Kit
Quantity 1 Kit
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.