Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gentian Violet, 96%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
125 g
1 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Gentian Violet , also known as crystal violet, is a triarylmethane dye that is used as a histological stain. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS Max % | 100% |
Percent Purity | 96% |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Color | Dark Green |
Quantity | 1 kg |
Chemical Name or Material | Gentian Violet |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction