Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Educational Insights GeoSafari™ Compass Binoculars

Click to view available options
Class Size:
1 Student
Description
- Bring outdoor expeditions into clearer focus with the GeoSafari Compass Binoculars
- Featuring 4x magnification, 30 mm glass lenses, an easy-focus knob, handy neck strap, and built-in compass, this set has everything junior explorers need to investigate the great outdoors or trips into town.
- GeoSafari Compass Binoculars are perfect for STEM learning.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | GeoSafari Compass Binoculars |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction