Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Educational Insights Geosafari™ Day 'N' Night Ant Factory
SCI_ED

Catalog No. S02732
Click to view available options
Class Size:
1 Class

Get a firsthand look at the amazing working world of ants with the Day 'N' Night Ant Farm. The translucent, nutrient-rich, farm gel doubles as habitat and food that ants can work with while they eat, eliminating the need for food or water

  • Get a firsthand look at the amazing working world of ants with the Day 'N' Night Ant Farm.
  • The translucent, nutrient-rich, farm gel doubles as habitat and food that ants can work with while they eat, eliminating the need for food or water.
  • Witness their amazing feats by day, then switch on the color-changing LED lights and watch as the ants work the nightshift.
  • Custom air vents keep ants safe and ventilation flowing throughout their workspace.
  • Includes an activity guide packed with awesome ant inspiration.

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Class
Product Type GeoSafari Ant Factories
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.