Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Educational Insights GeoSafari Jr. Talking Miroscope
SCI_ED

Catalog No. S02746
Click to view available options
Class Size:
1 Student

Zoom in on amazing images and learn fun facts about animals, plants, and interesting household items with the GeoSafari Jr. Talking Microscope! Twenty slides feature 60 incredible images. Wildlife conservationist Bindi Irwin voices facts and quiz questions

  • Zoom in on amazing images and learn fun facts about animals, plants, and interesting household items with the GeoSafari Jr. Talking Microscope
  • Twenty slides feature 60 incredible images.
  • Wildlife conservationist Bindi Irwin voices facts and quiz questions.
  • GeoSafari Jr. Talking Microscope requires 3 AAA batteries (not included).

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Student
Product Type Microscope
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.