Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Educational Insights GeoSafari™ Pocket Scope

Click to view available options
Class Size:
1 Student
Description
- See things in incredibly magnified detail, both up close and far away, with the GeoSafari Pocket Scope
- Study any object at 30x magnification with this 2-in-1 microscoping tool. Then extend the barrel to transform the scope into an 8x telescope.
- Bring nature up close with the GeoSafari Pocket Scope.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | GeoSafari Pocket Scope |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction