Learn More
Description
Includes
Fossil collection, igneous rock collection, metamorphic rock collection, mineral collection and sedimentary rock collection
Each collection features a selection of hand-picked specimens chosen for color, texture and beauty. Also contains a four-color identification chart, self-adhesive labels, handy storage tray and an information-packed guide.
Individual kits can be found at www.fishersci.edu.
Specifications
Specifications
| Grade | 3+ |
| For Use With (Application) | Dig into some serious fun with these fascinating collection kits |
| For Use With (Equipment) | Students 8 to 12yrs |
| Includes | 4 Color identification chart, self-adhesive labels, handy storage tray, and an information-packed guide, 9 fascinating fossils, 12 exotic rock specimens |
| Product Type | Igneous Rocks Collection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
