Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gephyrin/GPHN Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB268288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB268288 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB268288 Supplier Novus Biologicals Supplier No. NBP234859JF549
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gephyrin/GPHN Polyclonal antibody specifically detects Gephyrin/GPHN in Human samples. It is validated for Western Blot

Specifications

Antigen Gephyrin/GPHN
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias gephyrin, GPHRYN, KIAA1385GPHGEPH
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 686 to 736 of human Gephyrin using the numbering given in entry NP_001019389.1 (GeneID 10243).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10243
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.