Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gephyrin/GPHN Antibody (OTI3B6), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271556G
Description
Gephyrin/GPHN Monoclonal specifically detects Gephyrin/GPHN in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Gephyrin/GPHN | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GPHN | |
Full length recombinant protein of human Gephyrin/GPHN (NP_065857) produced in HEK293T cells. | |
0.1 mL | |
10243.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3B6 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
gephyrin, GPHRYN, KIAA1385GPHGEPH | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction