Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gephyrin/GPHN Antibody (OTI3B6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271556MFV610
Description
Gephyrin/GPHN Monoclonal antibody specifically detects Gephyrin/GPHN in Human, Mouse, Rat, Monkey samples. It is validated for Flow CytometrySpecifications
Gephyrin/GPHN | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Flow Cytometry | |
OTI3B6 | |
Flow Cytometry | |
gephyrin, GPHRYN, KIAA1385GPHGEPH | |
Full length human recombinant protein of human GPHN (NP_065857) produced in HEK293T cell. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Neuroscience | |
10243 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction