Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gephyrin Recombinant Rabbit Monoclonal Antibody (JG34-47)
GREENER_CHOICE

Catalog No. PIMA534727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534727 Supplier Invitrogen™ Supplier No. MA534727
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: SK-Br-3, rat kidney tissue, human kidney tissue, mouse brain tissue.

Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gephyrin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JG34-47
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Gphn
Gene Accession No. Q03555, Q8BUV3, Q9NQX3
Gene Alias 5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein
Gene Symbols Gphn
Host Species Rabbit
Immunogen Recombinant protein within Human Gephyrin aa 5-95.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10243, 268566, 64845
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.