Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Geraniol, 97%

Catalog No. aaa1373618
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 106-24-1 | C10H18O | 154.253 g/mol

Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

Solubility
Not miscible in water.

Notes
Incompatible with Strong oxidizing agents, acid chlorides, acid anhydrides. Keep away from heat. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-24-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00002917
InChI Key GLZPCOQZEFWAFX-JXMROGBWSA-N
Synonym geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
PubChem CID 637566
ChEBI CHEBI:17447
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(=CCCC(=CCO)C)C

Specifications

Melting Point -15°C
Density 0.884
Boiling Point 114°C to 115°C (12 mmHg)
Flash Point 101°C (213°F)
Odor Floral
Refractive Index 1.476
Quantity 50 g
Beilstein 1722456
Merck Index 14,4403
Solubility Information Not miscible in water.
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material Geraniol
Show More Show Less
Hazard Category H227-H315-H317-H318
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P370+P378q-P501c
EINECSNumber 203-377-1
RTECSNumber RG5830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.