Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluenesulfonic acid, pure, 12% in acetic acid

Catalog No. AC421220010
Click to view available options
Quantity:
250 g
1 kg

CAS: 104-15-4 | C7H8O3S | 172.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Toluenesulfonic acid
Color Colorless to Yellow
Name Note 12% in Acetic Acid
CAS 104-15-4, 64-19-7
Boiling Point 116.0°C
Molecular Formula C7H8O3S
MDL Number MFCD00064387
Beilstein 11, 97
Concentration 11 to 13% (Alkalimetry)
Fieser 01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Merck Index 15, 9692
Density 1.0700g/mL
Synonym p-toluenesulfonic acid, p-toluenesulphonic acid, 4-toluenesulfonic acid, tosic acid, p-tolylsulfonic acid, toluene-4-sulfonic acid, toluenesulfonic acid, tosylic acid, benzenesulfonic acid, 4-methyl, p-methylbenzenesulfonic acid
Solubility Information Solubility in water: soluble.
InChI Key JOXIMZWYDAKGHI-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)O
IUPAC Name 4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 172.198
PubChem CID 6101
ChEBI CHEBI:27849
Formula Weight 172.2
Grade Pure
Packaging Glass bottle
Refractive Index 1.3825 to 1.3845
Specific Gravity 1.07
Quantity 1 kg
Flash Point 41°C
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.