Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Germaine™ Laboratories Febrile Antigens For Salmonella H
Febrile antigens intended for detection of bacterial agglutinins associated with Salmonella H infection
$62.50
Catalog Number | Mfr. No. | Type | Detectable Analytes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Detectable Analytes | Price | Quantity & Availability | ||||
16-414-118
|
Germaine™ Laboratories
1401 |
Salmonella H b Antigen | Salmonella H B Antibodies |
Each for $62.50
|
|
||||
16-414-117
|
Germaine™ Laboratories
1400 |
Salmonella H a Antigen | Salmonella H A Antibodies |
Each for $62.50
|
|
||||
16-414-120
|
Germaine™ Laboratories
1403 |
Salmonella H d Antigen | Salmonella H D Antibodies |
Each for $62.50
|
|
||||
Description
- 5mL vial
Specifications
5 mL |
Professional use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title