Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Febrile Antigens For Salmonella O

Febrile Antigens intended for detection of bacterial agglutinins associated with Salmonella O infection

$63.30

Specifications

Quantity 5 mL
View More Specs

Products 2
Catalog Number Mfr. No. Type Detectable Analytes Price Quantity & Availability  
Catalog Number Mfr. No. Type Detectable Analytes Price Quantity & Availability  
16-414-101
View Documents
Germaine™ Laboratories
1501
Somatic Antigen Salmonella O Group B Agglutinins
Each for $63.30
Only null left
Add to Cart
 
16-414-103
View Documents
Germaine™ Laboratories
1503
Salmonella O Group D Antigen Salmonella O Group D Agglutinins
Each for $63.30
Only null left
Add to Cart
 
Description

Description

  • 5mL vial

Specifications

Specifications

5 mL
Videos
SDS
Documents

Documents

Product Certifications

Professional use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.